Learn More
High-Level Rare Earth Carbonatite, RM, ARMI∣MBH
Specifications
Specifications
| Chemical Name or Material | Aluminium∣Aluminium oxide∣Barium∣Barium oxide∣Calcium∣Calcium oxide∣Cerium∣Cerium(IV) oxide∣Dysprosium∣Europium∣Holmium∣Iron∣Iron (III) Oxide∣Lanthanum∣Lanthanum(III) Oxide∣Lead∣Loss on ignition∣Lutetium∣Magnesium∣Magnesium oxide∣Manganese∣Manganese(II) oxide∣Neodymium∣Neodymium(III) oxide∣Niobium∣Phosphorus∣Phosphorus pentoxide∣Potassium∣Potassium oxide∣Praseodymium∣Praseodymium oxide∣Rubidium∣Samarium∣Scandium∣Silicon∣Silicon dioxide∣Strontium∣Strontium oxide∣Sulfur∣Sulfur trioxide∣Thorium∣Titanium∣Titanium dioxide∣Total Light Rare Earth Element Oxides∣Total Light Rare Earth Elements∣Uranium∣Ytterbium∣Yttrium |
| Quantity | 90 g |
| Weight | 90 g |
| Form | Powder |
| Concentration or Composition (by Analyte or Components) | Titanium 0.012 %∣Titanium dioxide 0.021 %∣Potassium 0.093 %∣Potassium oxide 0.11 %∣Phosphorus 0.13 %∣Praseodymium 0.16 %∣Aluminium 0.18 %∣Manganese 0.18 %∣Praseodymium oxide 0.2 %∣Manganese(II) oxide 0.23 %∣Phosphorus pentoxide 0.3 %∣Aluminium oxide 0.33 %∣Neodymium 0.45 %∣Neodymium(III) oxide 0.52 %∣Lutetium 0.6 mg/kg∣Iron 0.93 %∣Iron (III) Oxide 1.3 %∣Lanthanum 1.3 %∣Lanthanum(III) Oxide 1.5 %∣Strontium 1.6 %∣Magnesium 1.8 %∣Cerium 1.9 %∣Strontium oxide 1.9 %∣Cerium(IV) oxide 2.3 %∣Sulfur 2.7 %∣Magnesium oxide 3 %∣Holmium 3.3 mg/kg∣Silicon 3.3 %∣Rubidium 3.8 mg/kg∣Total Light Rare Earth Elements 3.8 %∣Ytterbium 4.1 mg/kg∣Total Light Rare Earth Element Oxides 4.5 %∣Scandium 5.7 mg/kg∣Sulfur trioxide 6.6 %∣Silicon dioxide 7.2 %∣Barium 8.9 %∣Barium oxide 9.9 %∣Uranium 17 mg/kg∣Niobium 23 mg/kg∣Calcium 23.7 %∣Dysprosium 25 mg/kg∣Loss on ignition 30.4 %∣Calcium oxide 33.1 %∣Europium 50 mg/kg∣Yttrium 89 mg/kg∣Thorium 127 mg/kg∣Samarium 303 mg/kg∣Lead 1990 mg/kg |