missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hippocalcin Recombinant Rabbit Monoclonal Antibody (23GB6405), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553041
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of neuron-specific calcium-binding proteins family found in the retina and brain. This protein is associated with the plasma membrane. It has similarities to proteins located in the photoreceptor cells that regulate photosignal transduction in a calcium-sensitive manner. This protein displays recoverin activity and a calcium-dependent inhibition of rhodopsin kinase. It is identical to the rat and mouse hippocalcin proteins and thought to play an important role in neurons of the central nervous system in a number of species.
Specifications
| Hippocalcin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HPCA | |
| BDR2; calcium-binding protein; calcium-binding protein BDR-2; DYT2; hippocalcin; Hpca; hypothetical protein MGC73126; neuron-specific calcium-binding protein hippocalcin; P23K; zgc:73126 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15444, 29177, 3208 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6405 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P84074, P84075, P84076 | |
| HPCA | |
| A synthesized peptide derived from human Hippocalcin (1-36AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction