missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hippocalcin Recombinant Rabbit Monoclonal Antibody (23GB6405), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | Hippocalcin |
|---|---|
| Clone | 23GB6405 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088712
|
Invitrogen™
MA553040 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089699
|
Invitrogen™
MA553041 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hippocalcin Recombinant Monoclonal antibody specifically detects Hippocalcin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Hippocalcin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P84074, P84075, P84076 | |
| 15444, 29177, 3208 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6405 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BDR2; calcium-binding protein; calcium-binding protein BDR-2; DYT2; hippocalcin; Hpca; hypothetical protein MGC73126; neuron-specific calcium-binding protein hippocalcin; P23K; zgc:73126 | |
| A synthesized peptide derived from human Hippocalcin (1-36AA). | |
| Primary | |
| -20°C | |
| HPCA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title