Learn More
Invitrogen™ Hippocalcin Recombinant Rabbit Monoclonal Antibody (23GB6405), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553040
 View more versions of this product
Product Code. 30088712
  • € 446.00 / 100µL
Description
The protein encoded by this gene is a member of neuron-specific calcium-binding proteins family found in the retina and brain. This protein is associated with the plasma membrane. It has similarities to proteins located in the photoreceptor cells that regulate photosignal transduction in a calcium-sensitive manner. This protein displays recoverin activity and a calcium-dependent inhibition of rhodopsin kinase. It is identical to the rat and mouse hippocalcin proteins and thought to play an important role in neurons of the central nervous system in a number of species.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hippocalcin
Recombinant Monoclonal
Unconjugated
HPCA
BDR2; calcium-binding protein; calcium-binding protein BDR-2; DYT2; hippocalcin; Hpca; hypothetical protein MGC73126; neuron-specific calcium-binding protein hippocalcin; P23K; zgc:73126
Rabbit
Affinity chromatography
RUO
15444, 29177, 3208
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB6405
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P84074, P84075, P84076
HPCA
A synthesized peptide derived from human Hippocalcin (1-36AA).