missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2AD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11870-50ug
This item is not returnable.
View return policy
Description
HIST1H2AD Polyclonal antibody specifically detects HIST1H2AD in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP)
Specifications
| HIST1H2AD | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Polyclonal antibody raised in rabbit against HIST1H2AD using a KLH-conjugated synthetic peptide containing an unmodified sequence from the C-terminal part of the protein. | |
| 50 μg | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunofluorescence, ChIP Assay | |
| 4 mg/mL | |
| Western Blot 1:2000, ELISA 1:100-1:1000, Immunocytochemistry/ Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 1-2ug/ChIP | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction