missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2AD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 393.00 - € 549.00
Specifications
| Antigen | HIST1H2AD |
|---|---|
| Concentration | 4 mg/mL |
| Dilution | Western Blot 1:2000, ELISA 1:100-1:1000, Immunocytochemistry/ Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 1-2ug/ChIP |
| Applications | Western Blot, ELISA, Immunofluorescence, ChIP Assay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697527
|
Novus Biologicals
NBP3-11870-25ug |
25 μg |
€ 393.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627717
|
Novus Biologicals
NBP3-11870-50ug |
50 μg |
€ 549.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIST1H2AD Polyclonal antibody specifically detects HIST1H2AD in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP)Specifications
| HIST1H2AD | |
| Western Blot 1:2000, ELISA 1:100-1:1000, Immunocytochemistry/ Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 1-2ug/ChIP | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| 4 mg/mL | |
| Western Blot, ELISA, Immunofluorescence, ChIP Assay | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Polyclonal antibody raised in rabbit against HIST1H2AD using a KLH-conjugated synthetic peptide containing an unmodified sequence from the C-terminal part of the protein. | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title