missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine H2R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 409.50 - € 540.75
Specifications
| Antigen | Histamine H2R |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18487140
|
Novus Biologicals
NBP1-86082-25ul |
25 μL |
€ 433.00 € 409.50 / 25µL Save € 23.50 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18225436
|
Novus Biologicals
NBP1-86082 |
0.1 mL |
€ 572.00 € 540.75 / 0.10mL Save € 31.25 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histamine H2R Polyclonal specifically detects Histamine H2R in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Histamine H2R | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| gastric receptor 1, Gastric receptor I, H2R, HH2R, histamine H2 receptor, histamine receptor H2 | |
| HRH2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| GPCR, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 3274 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NRDFRTGYQQLFCCRLANRNSHKTSLRSNASQLSRTQSREPRQQEEKPLKLQVWSGTEVTAPQGATDR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title