missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B Antibody (SR1182), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21669-50ul
This item is not returnable.
View return policy
Description
Histone H2B Monoclonal antibody specifically detects Histone H2B in Yeast samples. It is validated for Western Blot
Specifications
| Histone H2B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Yeast | |
| Purified |
| Western Blot | |
| SR1182 | |
| Western Blot 1:500-1:2000 | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| A synthesized peptide derived from yeast Histone H2B (Uniprot #: P02293) | |
| 50 μg | |
| Chromatin Research, Epigenetics | |
| 440689 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction