missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3.3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | Histone H3.3A |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641436
|
Novus Biologicals
NBP2-98768-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606295
|
Novus Biologicals
NBP2-98768-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H3.3A Polyclonal antibody specifically detects Histone H3.3A in Human samples. It is validated for Western BlotSpecifications
| Histone H3.3A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Epigenetics | |
| PBS (pH 7.0) | |
| 3020 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Histone H3.3A. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title