missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 gp120 Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-41090-0.025mg
This item is not returnable.
View return policy
Description
HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| HIV-1 gp120 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| env | |
| Antibody was raised against 15 amino acids from near the center of gp120. The immunogen is located within amino acids 470 - 520 of gp120. Amino Acid Squence: APTKAKRRVVQREKR | |
| 0.025 mg | |
| Primary | |
| Virus | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA | |
| Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 155971 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction