missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 gp120 Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
€ 173.00 - € 505.00
Especificaciones
| Antigen | HIV-1 gp120 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Descripción
HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA.Especificaciones
| HIV-1 gp120 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 155971 | |
| Antibody was raised against 15 amino acids from near the center of gp120. The immunogen is located within amino acids 470 - 520 of gp120. Amino Acid Squence: APTKAKRRVVQREKR | |
| Primary |
| 1 mg/mL | |
| Polyclonal | |
| Goat | |
| Virus | |
| Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
| env | |
| IgG | |
| Affinity Purified |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto