missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLTF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38528-20ul
This item is not returnable.
View return policy
Description
HLTF Polyclonal antibody specifically detects HLTF in Human samples. It is validated for ELISA,Western Blot
Specifications
| HLTF | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DNA-binding protein/plasminogen activator inhibitor 1 regulator, EC 3.6.1, EC 3.6.4.-, EC 6.3.2.-, helicase-like transcription factor, HIP116ARING finger protein 80, HIP116matrix associated, actin dependent regulator of chromatin, RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3, subfamily a, member 3, Sucrose nonfermenting protein 2-like 3, ZBU1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 726-1009 of human HLTF (NP_620636.1).,, Sequence:, VSSNGPSGNDTPEELRKKLIRKMKLILSSGSDEECAICLDSLTVPVITHCAHVFCKPCICQVIQNEQPHAKCPLCRNDIHEDNLLECPPEELARDSEKKSDMEWTSSSKINALMHALTDLRKKNPNIKSLVVSQFTTFLSLIEIPLKASGFVFTRLDGSMAQKKRVESIQCFQNTEAGSPTIMLLSLKAGGVGLNLSAASRVFLMDPAWNPAAEDQCFDRCHRLGQKQEVIITKFIVKDSVEENMLKIQNKKRELAAGAFGTKKPNADEMKQAKINEIRTLIDL | |
| 20 μL | |
| Cancer, Cell Biology, Chromatin Research, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| 6596 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction