missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLTF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | HLTF |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230767
|
Novus Biologicals
NBP3-38528-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229415
|
Novus Biologicals
NBP3-38528-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLTF Polyclonal antibody specifically detects HLTF in Human samples. It is validated for ELISA,Western BlotSpecifications
| HLTF | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Chromatin Research, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 6596 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DNA-binding protein/plasminogen activator inhibitor 1 regulator, EC 3.6.1, EC 3.6.4.-, EC 6.3.2.-, helicase-like transcription factor, HIP116ARING finger protein 80, HIP116matrix associated, actin dependent regulator of chromatin, RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3, subfamily a, member 3, Sucrose nonfermenting protein 2-like 3, ZBU1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 726-1009 of human HLTF (NP_620636.1).,, Sequence:, VSSNGPSGNDTPEELRKKLIRKMKLILSSGSDEECAICLDSLTVPVITHCAHVFCKPCICQVIQNEQPHAKCPLCRNDIHEDNLLECPPEELARDSEKKSDMEWTSSSKINALMHALTDLRKKNPNIKSLVVSQFTTFLSLIEIPLKASGFVFTRLDGSMAQKKRVESIQCFQNTEAGSPTIMLLSLKAGGVGLNLSAASRVFLMDPAWNPAAEDQCFDRCHRLGQKQEVIITKFIVKDSVEENMLKIQNKKRELAAGAFGTKKPNADEMKQAKINEIRTLIDL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title