Learn More
HMG20B Mouse anti-Human, Unconjugated, Clone: 4C2A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67354-1-IG-150UL
Description
The HMG20B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HMG20B. This antibody recognizes human, mouse, rat antigen. The HMG20B antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| HMG20B | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 4C2A9 | |
| Unconjugated | |
| 150 μL | |
| 10362 | |
| IgG1 |