missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20B Mouse anti-Human, Unconjugated, Clone: 4C2A9, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | HMG20B |
|---|---|
| Clone | 4C2A9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16809945
|
Proteintech
67354-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16899935
|
Proteintech
67354-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HMG20B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HMG20B. This antibody recognizes human, mouse, rat antigen. The HMG20B antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| HMG20B | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 10362 | |
| Primary |
| 4C2A9 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title