Learn More
HMG20B Mouse anti-Human, Unconjugated, Clone: 4C2A9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67354-1-IG-150UL
 View more versions of this product
Product Code. 16899935
  • € 410.00 / 150µL
Description
The HMG20B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HMG20B. This antibody recognizes human, mouse, rat antigen. The HMG20B antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
HMG20B
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
4C2A9
Unconjugated
150 μL
10362