missing translation for 'onlineSavingsMsg'
Learn More

HMGB3/HMG4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 280.00 - € 624.00

Specifications

Antigen HMGB3/HMG4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18798313
View Documents
Novus Biologicals
NBP2-47434
0.1 mL
€ 624.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18656045
View Documents
Novus Biologicals
NBP2-47434-25ul
25 μL
€ 280.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HMGB3/HMG4 Polyclonal specifically detects HMGB3/HMG4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

HMGB3/HMG4
Polyclonal
Rabbit
Chromatin Research, DNA Repair
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
3149
This antibody was developed against a recombinant protein corresponding to amino acids: EKYEKDVADYKSKGKFDGAKGPAKVARKKVEEEDEE
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein
HMGB3
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.