missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP U Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 752.00
Specifications
| Antigen | hnRNP U |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666358
|
Novus Biologicals
NBP2-48729-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698678
|
Novus Biologicals
NBP2-48729 |
0.1 mL |
€ 752.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
hnRNP U Polyclonal antibody specifically detects hnRNP U in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| hnRNP U | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers | |
| PBS (pH 7.2), 40% Glycerol | |
| 3192 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| heterogeneous nuclear ribonucleoprotein U, heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A), hnRNP U, HNRPU, p120, p120 nuclear protein, pp120, SAFA, SAF-AhnRNPU, Scaffold attachment factor A, U21.1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TEQKGGDKKRGVKRPREDHGRGYFEYIEENKYSRAKSPQPPVEEEDEHFDDTVVCLDT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title