Learn More
Honeywell KCL™ Honeywell KCL™ Butoject™ 898 Gloves
Description
- AQL < 0.65 excellent low degradation manufactured by Vulkoject process gastight (meets NATO requirements)
- Loose fit allows gloves to be worn underneath
- Can be re-used after contamination with certain chemicals after special cleaning process (please consult KCL Laboratories)
- Can be combined with chemical protective clothing
Specifications
Specifications
| Certifications/Compliance | 951037 |
| Color | Black |
| Material | Butyl |
| Product Type | Gloves |
| Size | 11 |
| Thickness (Metric) | 0.7 cm |
| Lining | Heavy chemical |
| Length (Metric) | 340-360 cm |