Learn More
Honeywell KCL™ Honeywell KCL™ Butoject™ 898 Gloves
Product Code. 10329784
Change view
Click to view available options
:
Size: 11
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10329784 Size: 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10329784 Supplier Honeywell KCL™ Supplier No. 898/11
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Made from butyl rubber. Honeywell KCL™ Honeywell KCL™ Butoject™ 898 Gloves are manufactured with environmentally-friendly injection molding process. The glove is very popular in the field of NBC protection to its excellent protection against wide range of chemical and gases.
  • AQL < 0.65 excellent low degradation manufactured by Vulkoject process gastight (meets NATO requirements)
  • Loose fit allows gloves to be worn underneath
  • Can be re-used after contamination with certain chemicals after special cleaning process (please consult KCL Laboratories)
  • Can be combined with chemical protective clothing
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance 951037
Color Black
Material Butyl
Product Type Gloves
Size 11
Thickness (Metric) 0.7 cm
Lining Heavy chemical
Length (Metric) 340-360 cm