Learn More
Honeywell KCL™ Honeywell KCL™ K-Tex™ 930
Click to view available options
Size:
7
8
9
10
Unit Size:
10 pairs
Description
- Abrasion resistance
- Tear resistance
- Puncture resistancve
- Cutting resistance (Coupe test)
Specifications
Specifications
| Certifications/Compliance | 0075/1445/162/07/12/0519 |
| Color | Multicolored |
| Disposable | Single-use |
| Hand | Ambidextrous |
| Latex Free | No |
| Powdered | Powder Free |
| Size | 10 |
| Finger Style | Full fingers |
| Glove Cuff Color | Yellow |