Learn More
Honeywell KCL™ Honeywell KCL™ KarboTECT™ 950 Gloves
Click to view available options
Size:
9
10
Description
- Ambidextrous design makes them especially economical contact
- Heat up to 250 °C
- Suitable for food handling according to EU 1935/2004
- Good thermal insulation
Specifications
Certifications/Compliance | IFA 0401104 |
Color | Yellow, Black |
Latex Free | Yes |
Material | Para-Aramid, Carbon Fiber |
Powdered | Powder Free |
Product Type | Gloves |
Size | 9 |
Temperature (Metric) Max | 250°C |
Length (Metric) | 240-260 cm |