Learn More
Honeywell™ Leather with Cotton Heat Resistant Glove
Click to view available options
Size:
10
Conditionnement:
Each
Spécification
Spécification
| Certifications/Compliance | EC 0075/797/162/07/12/0583 |
| Classification | CE CATEGORY II |
| Material | Leather |
| Powdered | Powder Free |
| Product Type | Gloves |
| Size | 10 |
| Thickness (Metric) | 1.3 mm |
| Glove Cuff Style | Gauntlet |
| Liner Material | Cotton |
| Quantity | 1 Each |
| Afficher plus |