missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 227.00 - € 326.00
Specifications
| Antigen | HOXB8 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635745
|
Novus Biologicals
NBP2-99554-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690136
|
Novus Biologicals
NBP2-99554-50ul |
50 μg |
€ 227.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXB8 Polyclonal antibody specifically detects HOXB8 in Mouse samples. It is validated for Western BlotSpecifications
| HOXB8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| homeo box 2D, homeo box B8, homeobox B8, Homeobox protein Hox-2.4, Homeobox protein Hox-2D, homeobox protein Hox-B8, HOX2, HOX2DHox-2.4 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HOXB8 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 3218 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title