missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Antibody (SR1327), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21644-50ul
This item is not returnable.
View return policy
Description
HRAS Monoclonal antibody specifically detects HRAS in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Specifications
| HRAS | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| SR1327 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:30, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1 | |
| A synthesized peptide derived from human HRAS (Uniprot #: P01112) | |
| 50 μg | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| 3265 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction