missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Antibody (SR1327), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 373.00 - € 483.00
Specifications
| Antigen | HRAS |
|---|---|
| Clone | SR1327 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:30, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600565
|
Novus Biologicals
NBP3-21644-100ul |
100 μg |
€ 483.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699573
|
Novus Biologicals
NBP3-21644-50ul |
50 μg |
€ 373.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HRAS Monoclonal antibody specifically detects HRAS in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
| HRAS | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:30, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1 | |
| A synthesized peptide derived from human HRAS (Uniprot #: P01112) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1327 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 3265 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title