missing translation for 'onlineSavingsMsg'
Learn More

HSF1 Antibody (4B4), Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals NBP2-42206-0.025mg

 View more versions of this product

Product Code. 18696936

  • € 174.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HSF1 Monoclonal antibody specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSF1
Monoclonal
1 mg/mL
Western Blot 1:1000, ELISA, Immunohistochemistry 1:2000, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation, Gel Super Shift Assays
Q00613
Rat
Protein G purified
RUO
Primary
Human, Mouse, Rat, Bovine, Guinea Pig
Purified
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
4B4
Unconjugated
PBS (pH 7.4), 50% Glycerol
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Purified recombinant mouse HSF1 protein, epitope mapping to amino acids 425-439
0.025 mg
Cancer, DNA Repair, Transcription Factors and Regulators
3297
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.