missing translation for 'onlineSavingsMsg'
Learn More

HSF1 Antibody (4B4), Novus Biologicals™

Rat Monoclonal Antibody

€ 174.00 - € 579.00

Specifications

Antigen HSF1
Clone 4B4
Concentration 1 mg/mL
Dilution Western Blot 1:1000, ELISA, Immunohistochemistry 1:2000, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation, Gel Super Shift Assays
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18696936
View Documents
Novus Biologicals
NBP2-42206-0.025mg
0.025 mg
€ 174.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18640957
View Documents
Novus Biologicals
NBP2-42206
0.1 mg
€ 579.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HSF1 Monoclonal antibody specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays
Specifications

Specifications

HSF1
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Rat
Cancer, DNA Repair, Transcription Factors and Regulators
PBS (pH 7.4), 50% Glycerol
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Purified recombinant mouse HSF1 protein, epitope mapping to amino acids 425-439
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
4B4
Western Blot 1:1000, ELISA, Immunohistochemistry 1:2000, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation, Gel Super Shift Assays
Monoclonal
Purified
RUO
Human, Mouse, Rat, Bovine, Guinea Pig
Q00613
3297
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.