missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody (8A7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-96431-0.025mg
This item is not returnable.
View return policy
Description
HSP27 Monoclonal specifically detects HSP27 in Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Hamster, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| HSP27 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:5000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Knockout Validated | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects approx 25kDa or approx 27kDa. Recognizes HSP25 and HSP27, cross reacts with alpha B crystallin. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
| Western Blot | |
| 8A7 | |
| Unconjugated | |
| P04792 | |
| HSPB1 | |
| HSP27 peptide | |
| 0.025 mg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| 3315 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction