missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 Antibody (2D12), Novus Biologicals™
Rat Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-97450-0.05ML
This item is not returnable.
View return policy
Description
HSP90 Monoclonal specifically detects HSP90 in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Fungi, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| HSP90 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| HSP90AA1 | |
| Native mouse Hsp90. | |
| 0.05 mL | |
| Primary | |
| Reacts with Beluga, and water mold | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, Immunoprecipitation | |
| 2D12 | |
| Western Blot 1:500, Immunoprecipitation 1:10-1:500 | |
| FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| Rat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3320 | |
| Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Fungi, Guinea Pig, Hamster, Primate, Rabbit, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction