missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 Antibody (2D12), Novus Biologicals™
Rat Monoclonal Antibody has been used in 4 publications
€ 169.00 - € 695.00
Specifications
| Antigen | HSP90 |
|---|---|
| Clone | 2D12 |
| Dilution | Western Blot 1:500, Immunoprecipitation 1:10-1:500 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18033828
|
Novus Biologicals
NBP1-97450-0.025ML |
0.025 mL |
€ 169.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18069338
|
Novus Biologicals
NBP1-97450-0.05ML |
0.05 mL |
€ 388.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18094159
|
Novus Biologicals
NBP1-97450-0.2ML |
0.2 mL |
€ 695.00
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSP90 Monoclonal specifically detects HSP90 in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Fungi, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| HSP90 | |
| Western Blot 1:500, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 3320 | |
| Native mouse Hsp90. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 2D12 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rat | |
| Hamster, Rat, Rabbit, Primate, Chicken, Sheep, Mouse, Fish, Bovine, Guinea Pig, Human, Fungi, Canine | |
| FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| HSP90AA1 | |
| IgM | |
| Ammonium sulfate precipitation | |
| Reacts with Beluga, and water mold |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title