missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA8/HSC71/Hsc70 Antibody (3G10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26214-50ul
This item is not returnable.
View return policy
Description
HSPA8/HSC71/Hsc70 Monoclonal antibody specifically detects HSPA8/HSC71/Hsc70 in Human samples. It is validated for Western Blot, ELISA
Specifications
| HSPA8/HSC71/Hsc70 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3G10 | |
| Western Blot 1:500-1:5000, ELISA | |
| Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71 | |
| A synthesized peptide derived from Human HSPA8/HSC71/Hsc70 [UniProt P11142] | |
| 50 μL | |
| Membrane Trafficking and Chaperones | |
| 3312 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction