missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA8/HSC71/Hsc70 Antibody (3G10), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | HSPA8/HSC71/Hsc70 |
|---|---|
| Clone | 3G10 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30030542
|
Novus Biologicals
NBP3-26214-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30026883
|
Novus Biologicals
NBP3-26214-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSPA8/HSC71/Hsc70 Monoclonal antibody specifically detects HSPA8/HSC71/Hsc70 in Human samples. It is validated for Western Blot, ELISASpecifications
| HSPA8/HSC71/Hsc70 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71 | |
| A synthesized peptide derived from Human HSPA8/HSC71/Hsc70 [UniProt P11142] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3G10 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3312 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title