Learn More
HspB2 Antibody (4B9E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52490
 View more versions of this product
Product Code. 18286480
  • € 552.00 / 0.10mg
Description
HspB2 Monoclonal specifically detects HspB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HspB2
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
DMPK-binding protein, HspB2, MKBP
Mouse
20.2 kDa
0.1 mg
Primary
Human
Purified
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry
4B9E2
Unconjugated
PBS with 0.05% Sodium Azide
HSPB2
Purified recombinant fragment of human HSPB2 (AA: 1-182) expressed in E. Coli.
Protein A or G purified
RUO
3316
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.