missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspB2 Antibody (4B9E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | HspB2 |
|---|---|
| Clone | 4B9E2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18286480
|
Novus Biologicals
NBP2-52490 |
0.1 mg |
€ 552.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610988
|
Novus Biologicals
NBP2-52490-0.025mg |
0.025 mg |
€ 203.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HspB2 Monoclonal specifically detects HspB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| HspB2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| DMPK-binding protein, HspB2, MKBP | |
| HSPB2 | |
| IgG1 | |
| Protein A or G purified |
| 4B9E2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 3316 | |
| Purified recombinant fragment of human HSPB2 (AA: 1-182) expressed in E. Coli. | |
| Primary | |
| 20.2 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title