missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-2/CD86 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB10852-SP
This item is not returnable.
View return policy
Description
B7-2/CD86 Monoclonal antibody specifically detects B7-2/CD86 in Human samples. It is validated for ELISA
Specifications
| B7-2/CD86 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
| Human embryonic kidney cell HEK293-derived human B7-2/CD86, Leu26-Pro247, Accession # NP_787058 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 1036426 | |
| ELISA | |
| NP_787058 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 942 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction