missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-2/CD86 Antibody, R&D Systems™
Mouse Monoclonal Antibody
€ 201.00 - € 454.00
Specifications
| Antigen | B7-2/CD86 |
|---|---|
| Clone | 1036426 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18122621
|
R&D Systems
MAB10852-100 |
100 μg |
€ 454.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18102811
|
R&D Systems
MAB10852-SP |
25 μg |
€ 201.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B7-2/CD86 Monoclonal antibody specifically detects B7-2/CD86 in Human samples. It is validated for ELISASpecifications
| B7-2/CD86 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
| Human embryonic kidney cell HEK293-derived human B7-2/CD86, Leu26-Pro247, Accession # NP_787058 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1036426 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_787058 | |
| 942 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title