missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CKAP4/p63 Antibody, R&D Systems™
Mouse Monoclonal Antibody
€ 228.00 - € 526.00
Specifications
| Antigen | CKAP4/p63 |
|---|---|
| Clone | 1086526 |
| Dilution | Western Blot 1 μg/mL, Simple Western 4 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
| Applications | Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30245515
|
R&D Systems
MAB11600-SP |
25 μg |
€ 228.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30245507
|
R&D Systems
MAB11600-100 |
100 μg |
€ 526.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CKAP4/p63 Monoclonal antibody specifically detects CKAP4/p63 in Human samples. It is validated for Western Blot,Simple Western,Immunohistochemistry,ImmunocytochemistrySpecifications
| CKAP4/p63 | |
| Western Blot 1 μg/mL, Simple Western 4 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| 63 kDa membrane protein, CLIMP-63, cytoskeleton-associated protein 4, ERGIC-63, MGC99554, P63, transmembrane protein (63kD), endoplasmic reticulum/Golgi intermediatecompartment, type-II transmembrane protein p63 | |
| Mouse myeloma cell line, NS0-derived human CKAP-4, His128-Val602, Accession # Q07065 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1086526 | |
| Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q07065 | |
| 10970 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title