missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC17A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10147-100
This item is not returnable.
View return policy
Description
CLEC17A Monoclonal antibody specifically detects CLEC17A in Human samples. It is validated for Immunohistochemistry
Specifications
| CLEC17A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| C-type lectin domain family 17, member A, Prolectin | |
| CHO-derived human CLEC17A, Lys194-Cys378, Accession # Q6ZS10 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 1064709 | |
| Immunohistochemistry 5-15 μg/mL | |
| Q6ZS10 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 388512 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction