missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC17A Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 261.00 - € 583.00
Specifications
| Antigen | CLEC17A |
|---|---|
| Clone | 1064709 |
| Dilution | Immunohistochemistry 5-15 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18714534
![]() |
R&D Systems
MAB10147-SP |
25 μg |
€ 261.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744454
![]() |
R&D Systems
MAB10147-100 |
100 μg |
€ 583.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLEC17A Monoclonal antibody specifically detects CLEC17A in Human samples. It is validated for ImmunohistochemistrySpecifications
| CLEC17A | |
| Immunohistochemistry 5-15 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| C-type lectin domain family 17, member A, Prolectin | |
| CHO-derived human CLEC17A, Lys194-Cys378, Accession # Q6ZS10 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1064709 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6ZS10 | |
| 388512 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title