missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DNAM-1/CD226 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB10988-100
This item is not returnable.
View return policy
Description
DNAM-1/CD226 Monoclonal antibody specifically detects DNAM-1/CD226 in Human samples. It is validated for Flow Cytometry
Specifications
| DNAM-1/CD226 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
| Human embryonic kidney cell HEK293-derived human DNAM-1/CD226 protein, Glu19-Asn247, Accession # Q15762 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry | |
| 2770B | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Q15762 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 10666 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction