missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DNAM-1/CD226 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
€ 218.00 - € 492.00
Specifications
| Antigen | DNAM-1/CD226 |
|---|---|
| Clone | 2770B |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18109490
|
R&D Systems
MAB10988-SP |
25 μg |
€ 218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18152941
|
R&D Systems
MAB10988-100 |
100 μg |
€ 492.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAM-1/CD226 Monoclonal antibody specifically detects DNAM-1/CD226 in Human samples. It is validated for Flow CytometrySpecifications
| DNAM-1/CD226 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
| Human embryonic kidney cell HEK293-derived human DNAM-1/CD226 protein, Glu19-Asn247, Accession # Q15762 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2770B | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q15762 | |
| 10666 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title