Learn More
Human FGF-18 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6671-100
This item is not returnable.
View return policy
Description
FGF18 Monoclonal antibody specifically detects FGF18 in Human samples. It is validated for Immunohistochemistry
Specifications
| FGF18 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| FGF18, FGF-18ZFGF5, fibroblast growth factor 18, zFGF5 | |
| E. coli-derived human FGF-18 protein, Glu28-Ala207, Accession # O76093 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 91811 | |
| Immunohistochemistry 5-15 μg/mL | |
| O76093 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8817 | |
| Human | |
| Purified |