missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ig Kappa Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB100501-100
This item is not returnable.
View return policy
Description
Human Kappa Light Chain Monoclonal specifically detects Human Kappa Light Chain in Human samples. It is validated for Western Blot, Simple Western, ELISA.
Specifications
| Human Kappa Light Chain | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Human Ig Kappa light chain, | |
| 100 μg | |
| Detects Human Ig Kappa light chain in direct ELISAs. | |
| Human | |
| Supernatant |
| Western Blot, ELISA | |
| 999947 | |
| Western Blot 2 μg/mL, ELISA | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction