missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ig Kappa Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 206.00 - € 489.00
Specifications
| Antigen | Human Kappa Light Chain |
|---|---|
| Clone | 999947 |
| Dilution | Western Blot 2 μg/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18704357
![]() |
R&D Systems
MAB100501-SP |
25 μg |
€ 206.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18711117
![]() |
R&D Systems
MAB100501-100 |
100 μg |
€ 489.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human Kappa Light Chain Monoclonal specifically detects Human Kappa Light Chain in Human samples. It is validated for Western Blot, Simple Western, ELISA.Specifications
| Human Kappa Light Chain | |
| Western Blot 2 μg/mL, ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Human Ig Kappa light chain, | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Human Ig Kappa light chain in direct ELISAs. |
| 999947 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title