missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-33 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3625-SP
This item is not returnable.
View return policy
Description
IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| IL-33 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| O95760 | |
| IL33 | |
| E. coli-derived recombinant human IL-33 Ser112-Thr270 Accession # O95760 | |
| 25 μg | |
| Primary | |
| Detects human IL-33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-33 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 390412 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 90865 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction