missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-33 Antibody, R&D Systems™

Rat Monoclonal Antibody
€ 209.00 - € 516.00
Specifications
| Antigen | IL-33 |
|---|---|
| Clone | 390412 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18714819
![]() |
R&D Systems
MAB3625-SP |
25 μg |
€ 209.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18149270
![]() |
R&D Systems
MAB3625 |
100 μg |
€ 516.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| IL-33 | |
| LYOPH | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| O95760 | |
| 90865 | |
| E. coli-derived recombinant human IL-33 Ser112-Thr270 Accession # O95760 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-33 is observed. |
| 390412 | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
| IL33 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title