missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB4/CD85k/ILT3 Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB24252-100
This item is not returnable.
View return policy
Description
ILT3/CD85k/LILRB4 Monoclonal antibody specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| ILT3/CD85k/LILRB4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4 | |
| Mouse myeloma cell line, NS0-derived recombinant human LILRB4/CD85k/ILT3 protein, Pro17-His257, Accession # ABM86208 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry | |
| 1057706 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL | |
| ABM86208 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11006 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction