Learn More
Human NKp46/NCR1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB1850R-100
 View more versions of this product
Product Code. 18151021
  • € 651.00 / 100µg
Description
NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry, Agonist Activity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKp46/NCR1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Mouse T cell hybridoma transfected with human NKp46/NCR1, Accession # O76036
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
IgG2b
Flow Cytometry, Agonist Activity
195314R
Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity
O76036
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
9437