missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp46/NCR1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
€ 248.00 - € 651.00
Specifications
| Antigen | NKp46/NCR1 |
|---|---|
| Clone | 195314R |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity |
| Applications | Flow Cytometry, Agonist Activity |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18109200
|
R&D Systems
MAB1850R-SP |
25 μg |
€ 248.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18151021
|
R&D Systems
MAB1850R-100 |
100 μg |
€ 651.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry, Agonist ActivitySpecifications
| NKp46/NCR1 | |
| Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| Mouse T cell hybridoma transfected with human NKp46/NCR1, Accession # O76036 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 195314R | |
| Flow Cytometry, Agonist Activity | |
| Unconjugated | |
| Mouse | |
| Human | |
| O76036 | |
| 9437 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title