missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Prolactin Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11590-SP
This item is not returnable.
View return policy
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry,Western Blot,Simple Western
Specifications
| Prolactin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| PRL, prolactin | |
| E. coli- derived human Prolactin, Leu29-Cys227, Accession # P01236 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Simple Western | |
| 1085531 | |
| Western Blot 0.5 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL | |
| P01236 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5617 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction